Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic Anhydrous (Granular or Powder/Certified ACS), Fisher Chemical


Quantity 250 lb.
Packaging Fiber Drum

Manufacturer: fisher scientific company  FLS374250LB

 View more versions of this product

Catalog No. S374-250LB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99%
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Formula Weight 141.96
Quantity 250 lb.
pH 8.7 to 9.3
Molecular Formula HNa2O4P
Decomposition Information Decomposes information: > 240°C
Color White
Loss on Drying 0.2% max.
Heavy Metals (as Pb) 0.001% max.
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health:1 Flammability:0 Instability:1
Chemical Name or Material Sodium Phosphate Dibasic Anhydrous
MDL Number MFCD00003496
Sulfate 0.005% max.
Iron (Fe) 0.002% max.
Identification Pass Test
Synonym Disodium Hydrogen Phosphate
Assay Percent Range ≥99 %
Packaging Fiber Drum
Insoluble Matter 0.01% max.
Chloride 0.002% max.
Health Hazard 2 CAUTION!
ChemAlert Storage Symbol Gray
CAS 7558-79-4
Health Hazard 1 Signal Word:CAUTION!

Specifications

Chemical Name or Material Sodium Phosphate Dibasic Anhydrous
CAS 7558-79-4
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Assay ≥99%
Assay Percent Range ≥99 %
Molecular Formula HNa2O4P
Formula Weight 141.96
MDL Number MFCD00003496
Synonym Disodium Hydrogen Phosphate
Accessories

Accessories for Catalog Number: S374250LB

Item Mfr. No. Item Description Price Per Unit Quantity  
19-087809
Enpac™ Poly-Dolly™
spilkleen/cep
5300YE
Versatile all-in-one dolly, dispensing station and containment unit Each for $1,117.76
Accessories
 

Signal Word:CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health:1 Flammability:0 Instability:1

CAUTION!

SDS & Safety Documents