missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Phosphate, Dibasic, Anhydrous, BiotechGrade, 98-100.5%, Spectrum™ Chemical

S1401, 7558-79-4, Na2HPO4

$525.03 - $4,153.85

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
IUPAC Name disodium hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606729
View Documents Promotion Details
Spectrum Chemical Mfg Cor
S1401500GM
500 g Poly Bottle
Each for $525.03
 
18606730
View Documents Promotion Details
Spectrum Chemical Mfg Cor
S140125KG
2.5 kg Poly Bottle
Each for $1,695.47
 
18606731
View Documents Promotion Details
Spectrum Chemical Mfg Cor
S140112KG
12 kg Poly Pail
Each for $4,153.85
 
Description

Description

Spectrum™ Chemical Sodium Phosphate, Dibasic, Anhydrous, BiotechGrade is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Spectrum offers highly pure reagents suitable for biochemical research and analysis that are absent of inhibitors.

Specifications

Chemical Identifiers

7558-79-4
141.96
disodium hydrogen phosphate
HNa2O4P
BNIILDVGGAEEIG-UHFFFAOYSA-L
[Na+].[Na+].OP([O-])([O-])=O

Specifications

7558-79-4
500 g
HNa2O4P
[Na+].[Na+].OP([O-])([O-])=O
141.96
Biotech
0.0038
0.05
Poly Bottle
1.5e-005
1
None detected
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate
98.0 to 100.5%
1e-005
2e-005
Odorless
White powder
0.0019
SDS
Product Certifications