missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Phosphate, Dibasic, ACS Grade, ≥ 99.0%, LabChem™

$90.43 - $90.43

Chemical Identifiers

CAS 7558-79-4
CAS Max % 100
Linear Formula Na2HPO4
Formula Weight 141.96
Chemical Name or Material Sodium Phosphate, Dibasic
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC247741 Lab Chem
LC247741
500g Each for $90.43
Specifications Description & Specifications

Chemical Identifiers

7558-79-4
Na2HPO4
Sodium Phosphate, Dibasic
100
141.96
ACS Grade

Specifications

Crystalline Powder
7558-79-4
Passes Test
1.679g/cm3
Na2HPO4
100
Soluble in water
Sodium Phosphate, Dibasic
ACS Grade
141.96
1.679g/cm3
White
Poly Bottle
500g
Room Temperature
Phosphorus oxides
≥99.0%

Safety and Handling

GHS H Statement
Causes eye irritation.

GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 231-448-7