missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals AC204851000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysisSpecifications
Sodium phosphate, dibasic | |
7558-79-4 | |
8.7 to 9.3 (5% soln. at 20°C) | |
100 g | |
HNa2O4P | |
MFCD00003496 | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
disodium hydrogen phosphate | |
24203 | |
141.96 | |
Analytical | |
Plastic bottle |
p.a. | |
White | |
Granular Powder | |
99+% | |
Na2HPO4 | |
15, 8789 | |
Solubility in water: 75g/L (20°C). Other solubilities: practically insoluble in alcohol | |
[Na+].[Na+].OP([O-])([O-])=O | |
141.96 | |
CHEBI:34683 | |
≥99% | |
0.2% max. |
Chemical Identifiers
7558-79-4 | |
141.96 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
24203 | |
[Na+].[Na+].OP([O-])([O-])=O |
HNa2O4P | |
MFCD00003496 | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
CHEBI:34683 |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 231-448-7
RTECSNumber : WC4500000
TSCA : TSCA
RUO – Research Use Only