Please login to your online account to display your discounted pricing

Sodium phosphate, dibasic, +99%, for analysis, anhydrous, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  204855000

 View more versions of this product

Catalog No. AC204855000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 141.96
Physical Form Granular Powder
Quantity 500g
Color White
Merck Index 15, 8789
TSCA TSCA
RTECS Number WC4500000
EINECS Number 231-448-7
Purity Grade For analysis
pH 8.7 to 9.3 (5% soln. at 20°C)
Sulfate 50ppm max.
Potassium (K) 200ppm max.
Chlorine (Cl) 10ppm max.
Arsenic (As) 3ppm max.
Health Hazard 1 HYGROSCOPIC
Name Note p.a.
MDL Number MFCD00003496
Assay Percent Range 99+%
Linear Formula Na2HPO4
Chemical Name or Material Sodium phosphate, dibasic
CAS 7558-79-4
Molecular Formula HNa2O4P
Packaging Plastic bottle
Iron (Fe) 10ppm max.
Heavy Metals (as Pb) 10ppm max.
Synonym Disodium hydrogen phosphate
Insoluble Matter 0.01% max.
Loss on Drying 0.2% max.

Specifications

Chemical Name or Material Sodium phosphate, dibasic
Name Note p.a.
CAS 7558-79-4
Purity Grade For analysis
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula HNa2O4P
Linear Formula Na2HPO4
Formula Weight 141.96
MDL Number MFCD00003496
Synonym Disodium hydrogen phosphate
Accessories

HYGROSCOPIC

SDS & Safety Documents