Please login to your online account to display your discounted pricing

Sodium Phosphate ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Phosphate
Name Note Dibasic, Heptahydrate
CAS 7782-85-6
Purity Grade ACS
Molecular Formula Na2HPO4·7H2O
Formula Weight 268.07g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK7914500

 
avantor performance
791404
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging POLYSTORMOR
Quantity 500g
Chemical Name or Material Sodium Phosphate
Name Note Dibasic, Heptahydrate
CAS 7782-85-6
Purity Grade ACS
Molecular Formula Na2HPO4·7H2O
Formula Weight 268.07g/mol