Please login to your online account to display your discounted pricing

Sodium Phosphate ACS AR, Macron Fine Chemicals™


macron

Quantity 500g
Packaging POLYSTORMOR

Manufacturer: avantor performance  791404

 View more versions of this product

Catalog No. MK7914500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging POLYSTORMOR
Quantity 500g
Chemical Name or Material Sodium Phosphate
Name Note Dibasic, Heptahydrate
CAS 7782-85-6
Purity Grade ACS
Molecular Formula Na2HPO4·7H2O
Formula Weight 268.07g/mol

Specifications

Chemical Name or Material Sodium Phosphate
Name Note Dibasic, Heptahydrate
CAS 7782-85-6
Purity Grade ACS
Molecular Formula Na2HPO4·7H2O
Formula Weight 268.07g/mol