Please login to your online account to display your discounted pricing

Sodium Phosphate ACS AR Granular, Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Poly

Manufacturer: avantor performance  789206

 View more versions of this product

Catalog No. MK7892212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Poly
Quantity 2.5kg
Chemical Name or Material Sodium Phosphate
Name Note Monobasic, Monohydrate, Anhydrous
CAS 7558-79-4
Purity Grade ACS
Physical Form Granular
Molecular Formula Na2HPO4
Formula Weight 141.96g/mol

Specifications

Chemical Name or Material Sodium Phosphate
Name Note Monobasic, Monohydrate, Anhydrous
CAS 7558-79-4
Purity Grade ACS
Physical Form Granular
Molecular Formula Na2HPO4
Formula Weight 141.96g/mol