Please login to your online account to display your discounted pricing

Sodium Phosphate ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Phosphate
Name Note Monobasic, Monohydrate, Anhydrous
CAS 7558-79-4
Purity Grade ACS Grade
Physical Form Granular
Molecular Formula Na2HPO4
Formula Weight 141.96
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK7917212

 
avantor performance
791706
Glass 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 141.96
Physical Form Granular
Molecular Formula Na2HPO4
CAS 7558-79-4
Name Note Monobasic, Monohydrate, Anhydrous
Chemical Name or Material Sodium Phosphate
Packaging Glass
Quantity 2.5kg