Please login to your online account to display your discounted pricing

Sodium Phosphate ACS AR Granular, Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Glass

Manufacturer: avantor performance  791706

 View more versions of this product

Catalog No. MK7917212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Grade
Formula Weight 141.96
Physical Form Granular
Molecular Formula Na2HPO4
CAS 7558-79-4
Name Note Monobasic, Monohydrate, Anhydrous
Chemical Name or Material Sodium Phosphate
Packaging Glass
Quantity 2.5kg

Specifications

Chemical Name or Material Sodium Phosphate
Name Note Monobasic, Monohydrate, Anhydrous
CAS 7558-79-4
Purity Grade ACS Grade
Physical Form Granular
Molecular Formula Na2HPO4
Formula Weight 141.96