Please login to your online account to display your discounted pricing

Sodium Phosphate ACS AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Phosphate
Name Note Tribasic, 12-Hydrate
CAS 10101-89-0
Purity Grade ACS Grade
Physical Form Crystalline
Molecular Formula Na3PO4·12H2O
Formula Weight 380.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK7940500

 
avantor performance
794004
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 380.12
Chemical Name or Material Sodium Phosphate
Name Note Tribasic, 12-Hydrate
Packaging POLYSTORMOR
Physical Form Crystalline
Molecular Formula Na3PO4·12H2O
CAS 10101-89-0
Quantity 500g