Please login to your online account to display your discounted pricing

Sodium Phosphate ACS AR Crystal, Macron Fine Chemicals™


macron

Quantity 500g
Packaging POLYSTORMOR

Manufacturer: avantor performance  794004

 View more versions of this product

Catalog No. MK7940500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Grade
Formula Weight 380.12
Chemical Name or Material Sodium Phosphate
Name Note Tribasic, 12-Hydrate
Packaging POLYSTORMOR
Physical Form Crystalline
Molecular Formula Na3PO4·12H2O
CAS 10101-89-0
Quantity 500g

Specifications

Chemical Name or Material Sodium Phosphate
Name Note Tribasic, 12-Hydrate
CAS 10101-89-0
Purity Grade ACS Grade
Physical Form Crystalline
Molecular Formula Na3PO4·12H2O
Formula Weight 380.12