Please login to your online account to display your discounted pricing

Sodium Phosphate ACS AR Crystal, Macron Fine Chemicals™


macron

Quantity 12kg
Packaging STAKMOR

Manufacturer: avantor performance  794020

 View more versions of this product

Catalog No. MK794012

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging STAKMOR
Quantity 12kg
Chemical Name or Material Sodium Phosphate
Name Note Tribasic, 12-Hydrate
CAS 10101-89-0
Purity Grade ACS
Physical Form Crystalline
Molecular Formula Na3PO4·12H2O
Formula Weight 380.12g/mol

Specifications

Chemical Name or Material Sodium Phosphate
Name Note Tribasic, 12-Hydrate
CAS 10101-89-0
Purity Grade ACS
Physical Form Crystalline
Molecular Formula Na3PO4·12H2O
Formula Weight 380.12g/mol