missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium phosphate, 0.5M buffer soln., pH 9.0

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
MDL Number MFCD00146206
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60651AP View Documents Alfa Aesar™
J60651AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ60651AK View Documents Alfa Aesar™
J60651AK
250mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

Sodium phosphate buffer soln.
MFCD00146206