missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium phosphate, 0.5M buffer soln., pH 7.5

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
MDL Number MFCD00146206
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61561AK View Documents Alfa Aesar™
J61561AK
250mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ61561AP missing translation for 'viewDocuments' Alfa Aesar™
J61561AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Applications
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Solubility
Soluble in water (121 mg/ml at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong acids.
Specifications

Specifications

Sodium phosphate buffer soln.
MFCD00146206