Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium phosphate, 0.2M buffer soln., pH 8.0, low endotoxin

Chemical Identifiers

Chemical Name or Material Sodium phosphate
Name Note low endotoxin
MDL Number MFCD00146206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67215AE

 
alfa aesar™
J67215AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67215AK

 
alfa aesar™
J67215AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium phosphate
MDL Number MFCD00146206
pH 8.0
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid
Concentration 0.2M buffer soln.