missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium phosphate, 0.2M buffer soln., pH 7.6

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
MDL Number MFCD00146206
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62815AP View Documents Alfa Aesar™
J62815AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ62815K2 View Documents Alfa Aesar™
J62815K2
1L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

Sodium phosphate buffer soln.
MFCD00146206