Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium phosphate, 0.2M buffer soln., pH 7.5, low endotoxin

Chemical Identifiers

Chemical Name or Material Sodium phosphate
Name Note low endotoxin
MDL Number MFCD00146206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67331AE

 
alfa aesar™
J67331AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67331AK

 
alfa aesar™
J67331AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium phosphate
Physical Form Liquid
MDL Number MFCD00146206
pH 7.5
TSCA TSCA
Name Note low endotoxin
Concentration 0.2M buffer soln.
Color Colorless
Quantity 100mL