missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium phosphate, 0.2M buffer soln., pH 7.5

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
MDL Number MFCD00146206
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62041AP View Documents Alfa Aesar™
J62041AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ62041K2 View Documents Alfa Aesar™
J62041K2
1L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Applications
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately. It is also used in the preparation of mobile phase in high performance liquid chromatography (HPLC).

Solubility
Miscible with water. Immiscible with alcohol.

Notes
Incompatible with strong acids.
Specifications

Specifications

Sodium phosphate buffer soln.
MFCD00146206