Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium phosphate, 0.2M buffer soln., pH 7.2, low endotoxin


Quantity 100mL

Manufacturer: alfa aesar™  J67394AE

 View more versions of this product

Catalog No. AAJ67394AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

MDL Number MFCD00146206
Chemical Name or Material Sodium phosphate
pH 7.2
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid
Concentration 0.2M buffer soln.

Specifications

Chemical Name or Material Sodium phosphate
Name Note low endotoxin
MDL Number MFCD00146206