Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium phosphate, 0.2M buffer soln., pH 7.2, low endotoxin

Chemical Identifiers

Chemical Name or Material Sodium phosphate
Name Note low endotoxin
MDL Number MFCD00146206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67394AE

 
alfa aesar™
J67394AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67394AK

 
alfa aesar™
J67394AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00146206
Physical Form Liquid
Name Note low endotoxin
pH 7.2
TSCA TSCA
Quantity 100mL
Concentration 0.2M buffer soln.
Chemical Name or Material Sodium phosphate
Color Colorless