Learn More
Sodium persulfate, +98%, Thermo Scientific Chemicals
$81.97 - $319.38
Chemical Identifiers
CAS | 7775-27-1 |
---|---|
Molecular Formula | Na2O8S2 |
Molecular Weight (g/mol) | 238.092 |
MDL Number | MFCD00003501 |
InChI Key | CHQMHPLRPQMAMX-UHFFFAOYSA-L |
Synonym | sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate |
PubChem CID | 62655 |
IUPAC Name | disodium;sulfonatooxy sulfate |
SMILES | [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC202025000
|
Thermo Scientific Chemicals
202025000 |
500 g | Plastic bottle |
Each for $81.97
|
|
||||
AC202020010
|
Thermo Scientific Chemicals
202020010 |
1 kg | Plastic bottle |
Each for $128.89
|
|
||||
AC202020050
|
Thermo Scientific Chemicals
202020050 |
5 kg | Plastic bottle |
Each for $319.38
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7775-27-1 | |
238.092 | |
CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
62655 | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] |
Na2O8S2 | |
MFCD00003501 | |
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate | |
disodium;sulfonatooxy sulfate |
Specifications
7775-27-1 | |
Crystalline Powder and/or Crystals | |
98+% | |
Na2S2O8 | |
01,1102; 03,267 | |
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] | |
238.092 | |
238.09 | |
Pass Test | |
Sodium persulfate, 98+% |
White to Yellow | |
500 g | |
Na2O8S2 | |
MFCD00003501 | |
15, 8787 | |
CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
disodium;sulfonatooxy sulfate | |
62655 | |
≥98.0% (Iodometry) | |
Plastic bottle |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause sensitization by inhalation and skin contact.
Danger
EINECSNumber : 231-892-1
RTECSNumber : SE0525000
TSCA : TSCA
RUO – Research Use Only