missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium persulfate, +98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 202020050

 View more versions of this product

Catalog No. AC202020050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

Sodium persulfate
7775-27-1
Crystalline Powder and/or Crystals
98+%
Na2S2O8
01,1102; 03,267
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]
238.092
238.09
Pass Test
98+%
White to Yellow
5 kg
Na2O8S2
MFCD00003501
15, 8787
CHQMHPLRPQMAMX-UHFFFAOYSA-L
disodium;sulfonatooxy sulfate
62655
≥98.0% (Iodometry)
Plastic bottle

Chemical Identifiers

7775-27-1
238.092
CHQMHPLRPQMAMX-UHFFFAOYSA-L
62655
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]
Na2O8S2
MFCD00003501
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
disodium;sulfonatooxy sulfate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause sensitization by inhalation and skin contact.

Danger

EINECSNumber : 231-892-1

RTECSNumber : SE0525000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only