Please login to your online account to display your discounted pricing

Sodium Persulfate, 1.50 Molar, Ricca Chemical


Quantity 4L
Packaging Amber Plastic Bottle

Manufacturer: ricca chemical company  RIR75160004BV

 View more versions of this product

Catalog No. R75160004BV

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Sodium Persulfate
CAS Min % 72.0
CAS Max % 75.0
CAS 7732-18-5
CAS 7775-27-1
Packaging Amber Plastic Bottle
CAS Min % 25.8
CAS Max % 26.86

Specifications

Chemical Name or Material Sodium Persulfate
CAS 7732-18-5
CAS Min % 72.0
CAS Max % 75.0
CAS 7775-27-1
CAS Min % 25.8
CAS Max % 26.86
Physical Form Liquid