Please login to your online account to display your discounted pricing

Sodium Persulfate, 1.50 Molar, Ricca Chemical


Quantity 10L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR751600010F

 View more versions of this product

Catalog No. R751600010F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear colorless
Physical Form Liquid
Quantity 10L
Chemical Name or Material Sodium Persulfate
CAS 7732-18-5
Concentration or Composition Notes 1.50Molar
CAS Min % 72.0
CAS Max % 75.0
CAS 7775-27-1
Packaging Cubitainer
CAS Min % 25.8
CAS Max % 26.86

Specifications

Chemical Name or Material Sodium Persulfate
CAS 7732-18-5
CAS Min % 72.0
CAS Max % 75.0
CAS 7775-27-1
CAS Min % 25.8
CAS Max % 26.86
Physical Form Liquid