Please login to your online account to display your discounted pricing

Sodium Persulfate, 1.00 Molar, Ricca Chemical


Quantity 10L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR751500010F

 View more versions of this product

Catalog No. R751500010F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 10L
Chemical Name or Material Sodium Persulfate
CAS 7775-27-1
Assay Percent Range 1.000 ± 0.005 M at 20°C
Solvent or Matrix Aqueous Solution
CAS 7732-18-5
Packaging Cubitainer
Solubility Solubility in Water: Infinite
Odor Odorless
Name Note 1.00 Molar

Specifications

Chemical Name or Material Sodium Persulfate
Name Note 1.00 Molar
CAS 7775-27-1
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 1.000 ± 0.005 M at 20°C