missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium perrhenate, >99% (metals basis), Re 68% , Alfa Aesar™

Manufacturer:  Alfa Aesar 1141218

 View more versions of this product

Catalog No. AA1141218



Specifications

Specifications

Sodium perrhenate
13472-33-8
NaO4Re
White crystalline powder
KMISVFTVBOPTAI-UHFFFAOYSA-N
sodium;oxido(trioxo)rhenium
5107658
Crystalline
Yellow
Re 68%
99+% (metals basis)
MFCD00014231
sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium
[O-][Re](=O)(=O)=O.[Na+]
273.193
273.19
Odorless
50g

Chemical Identifiers

13472-33-8
273.193
KMISVFTVBOPTAI-UHFFFAOYSA-N
5107658
[O-][Re](=O)(=O)=O.[Na+]
NaO4Re
MFCD00014231
sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium
sodium;oxido(trioxo)rhenium
Safety and Handling

Safety and Handling

H272-H315-H319: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation.

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 236-742-9

Documents