Learn More
Sodium perrhenate, 99+% (metals basis), Re 68%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01141209
Description
Sodium perrhenate is used as a precursor to other rhenium compounds. Further, it reacts with sodium in ethanol to prepare nonahydridorhenate and with polysulfide solutions to get tetrathiorhenate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium perrhenate | |
Re 68% | |
Yellow | |
10 g | |
NaO4Re | |
sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium | |
KMISVFTVBOPTAI-UHFFFAOYSA-N | |
sodium;oxido(trioxo)rhenium | |
5107658 | |
Odorless | |
White crystalline powder |
300°C | |
13472-33-8 | |
Crystalline | |
99+% (metals basis) | |
MFCD00014231 | |
Soluble in water and ethanol. | |
[O-][Re](=O)(=O)=O.[Na+] | |
273.193 | |
273.19 | |
Hygroscopic | |
5.39 g/cm3 at 20°C |
Chemical Identifiers
13472-33-8 | |
273.193 | |
KMISVFTVBOPTAI-UHFFFAOYSA-N | |
5107658 | |
[O-][Re](=O)(=O)=O.[Na+] |
NaO4Re | |
MFCD00014231 | |
sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium | |
sodium;oxido(trioxo)rhenium |
Safety and Handling
P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H272-H315-H319
DOTInformation : DOT Class: 5.1, Packaging Group: II
EINECSNumber : 236-742-9
RTECSNumber : WD3675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only