missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium perrhenate, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 6210706

 View more versions of this product

Catalog No. AA6210706



Specifications

Specifications

Rhenium(VI) oxide
99.9% (metals basis)
MFCD00016306
YSZJKUDBYALHQE-UHFFFAOYSA-N
trioxorhenium
102110
Powder
Odorless
5g
1314-28-9
O3Re
rhenium trioxide, rhenium oxide reo3, rhenium vi oxide, unii-fet0y2c413, rhenia, reo3, rhenium vi oxide, rhenium vi oxide trace metals basis 1g
O=[Re](=O)=O
234.204
236.2
6.9 to 7.4g/cm3 at 20°C
Violet

Chemical Identifiers

1314-28-9
234.204
YSZJKUDBYALHQE-UHFFFAOYSA-N
102110
O=[Re](=O)=O
O3Re
MFCD00016306
rhenium trioxide, rhenium oxide reo3, rhenium vi oxide, unii-fet0y2c413, rhenia, reo3, rhenium vi oxide, rhenium vi oxide trace metals basis 1g
trioxorhenium
Safety and Handling

Safety and Handling

EINECSNumber : 215-228-8

SDS Documents