Please login to your online account to display your discounted pricing

Sodium peroxide, 96%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  207705000

 View more versions of this product

Catalog No. AC207705000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Granules
Formula Weight 77.97
Quantity 500g
Color Pale yellow
Merck Index 15, 8786
EINECS Number 215-209-4
TSCA TSCA
Density 2.8g/mL
Fieser 06,548; 07,341; 11,492
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.
CAS 1313-60-6
Linear Formula Na2O2
Chemical Name or Material Sodium peroxide
MDL Number MFCD00003497
Assay Percent Range 96%
Name Note p.a.
Molecular Formula Na2O2
Iron (Fe) 50ppm max.
Lead (Pb) 50ppm max.
Sulfate 50ppm max.
Chlorine (Cl) 50ppm max.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Sodium peroxide
Name Note p.a.
CAS 1313-60-6
Assay ≥95%
Assay Percent Range 96%
Molecular Formula Na2O2
Linear Formula Na2O2
Formula Weight 77.97
MDL Number MFCD00003497
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.

SDS & Safety Documents