missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium perfluorooctanoate, 97%

$82.50 - $325.39

Chemical Identifiers

CAS 335-95-5
Molecular Formula C8F15NaO2
Molecular Weight (g/mol) 436.052
MDL Number MFCD00040394
InChI Key LWHQXUODFPPQTL-UHFFFAOYSA-M
Synonym sodium perfluorooctanoate, sodium pentadecafluorooctanoate, sodium perfluorocaprylate, octanoic acid, pentadecafluoro-, sodium salt, perfluorooctanoic acid sodium salt, unii-63a7smf12m, pentadecafluorooctanoic acid, sodium salt, sodium pentadecafluoro octanoate, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, sodium salt 1:1, potassium pentadecafluorooctanoate
PubChem CID 23667657
IUPAC Name sodium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES C(=O)(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1698806 View Documents Alfa Aesar™
L1698806
5g Each for $82.50
 
AAL1698814 View Documents Alfa Aesar™
L1698814*
25g Each for $325.39
 
Description

Description

Applications
Sodium perfluorooctanoate on gel-to-liquid-crystalline phase transition of dipalmitoylphosphatidylcholine vesicle membrane.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

335-95-5
436.052
LWHQXUODFPPQTL-UHFFFAOYSA-M
23667657
C(=O)(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)[O-].[Na+]
C8F15NaO2
MFCD00040394
sodium perfluorooctanoate, sodium pentadecafluorooctanoate, sodium perfluorocaprylate, octanoic acid, pentadecafluoro-, sodium salt, perfluorooctanoic acid sodium salt, unii-63a7smf12m, pentadecafluorooctanoic acid, sodium salt, sodium pentadecafluoro octanoate, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, sodium salt 1:1, potassium pentadecafluorooctanoate
sodium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate

Specifications

~275°C
335-95-5
CF3(CF2)6CO2Na
sodium perfluorooctanoate, sodium pentadecafluorooctanoate, sodium perfluorocaprylate, octanoic acid, pentadecafluoro-, sodium salt, perfluorooctanoic acid sodium salt, unii-63a7smf12m, pentadecafluorooctanoic acid, sodium salt, sodium pentadecafluoro octanoate, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, sodium salt 1:1, potassium pentadecafluorooctanoate
C(=O)(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)[O-].[Na+]
436.052
436.05
Sodium perfluorooctanoate
5g
C8F15NaO2
MFCD00040394
LWHQXUODFPPQTL-UHFFFAOYSA-M
sodium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
23667657
97%
Safety and Handling

Safety and Handling

TSCA : TSCA