missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Perchlorate Monohydrate (Crystalline/Laboratory), Fisher Chemical

Manufacturer:  Thermo Scientific S360500

 View more versions of this product

Catalog No. S360-500




Specifications

Specifications

Sodium Perchlorate Monohydrate
ClH2NaO5
acmc-1bii1
O.[O-]Cl(=O)(=O)=O.[Na+]
140.45
140.46
Laboratory
Pass Test
White
6.0 to 8.0
7791-07-3
MFCD00149164
IXGNPUSUVRTQGW-UHFFFAOYSA-M
sodium;perchlorate;hydrate
516899
Solid
2.02g/cm³
Glass Bottle
130°C
500g

Chemical Identifiers

7791-07-3
140.45
IXGNPUSUVRTQGW-UHFFFAOYSA-M
516899
O.[O-]Cl(=O)(=O)=O.[Na+]
ClH2NaO5
MFCD00149164
acmc-1bii1
sodium;perchlorate;hydrate
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents