missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium perchlorate monohydrate, 97+%

Manufacturer:  Alfa Aesar™ A112430B

 View more versions of this product

Catalog No. AAA112430B


 

Description

Description

Applications
Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.

Solubility
Soluble in water.

Notes
It is a hygroscopic solid. Store under inert atmosphere and keep away from ignition sources.
Specifications

Specifications

Sodium perchlorate monohydrate
≥97%
MFCD00149164
14,8653
O.[O-]Cl(=O)(=O)=O.[Na+]
140.45
140.48 (122.46 Anhydrous)
130°C (decomposition)
7791-07-3
ClH2NaO5
UN1502
IXGNPUSUVRTQGW-UHFFFAOYSA-M
sodium;perchlorate;hydrate
516899
Odorless
1000g

Chemical Identifiers

7791-07-3
140.45
IXGNPUSUVRTQGW-UHFFFAOYSA-M
ClH2NaO5
MFCD00149164
516899
Safety and Handling

Safety and Handling

GHS H Statement
H271-H302
May cause fire or explosion
strong oxidizer.
Harmful if swallowed.

GHS P Statement
P221-P283-P210-P306+P360-P371+P380+P375-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the risk of explosion.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: SODIUM PERCHLORATE

RTECSNumber : SC9850000

TSCA : TSCA

SDS Documents