Learn More
Sodium perchlorate monohydrate, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A112430I
Description
Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.
Solubility
Soluble in water.
Notes
It is a hygroscopic solid. Store under inert atmosphere and keep away from ignition sources.
Specifications
Sodium perchlorate monohydrate | |
7791-07-3 | |
ClH2NaO5 | |
UN1502 | |
sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate | |
IXGNPUSUVRTQGW-UHFFFAOYSA-M | |
sodium;perchlorate;hydrate | |
516899 | |
≥97% | |
Hygroscopic |
130°C (decomposition) | |
5000 g | |
MFCD00149164 | |
14,8653 | |
Soluble in water. | |
O.[O-]Cl(=O)(=O)=O.[Na+] | |
140.45 | |
140.48 (122.46 Anhydrous) | |
Odorless | |
2.02 |
Chemical Identifiers
7791-07-3 | |
140.45 | |
IXGNPUSUVRTQGW-UHFFFAOYSA-M | |
516899 | |
O.[O-]Cl(=O)(=O)=O.[Na+] |
ClH2NaO5 | |
MFCD00149164 | |
sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate | |
sodium;perchlorate;hydrate |
Safety and Handling
GHS H Statement
H271-H302
May cause fire or explosion
strong oxidizer.
Harmful if swallowed.
P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P305+P351+P338-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c
H271-H302-H319-H373
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: SODIUM PERCHLORATE
EINECSNumber : 231-511-9
RTECSNumber : SC9850000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only