missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium perchlorate, anhydrous, ACS, 98.0 to 102.0%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1162322

 View more versions of this product

Catalog No. AA1162322



Specifications

Specifications

Sodium perchlorate, anhydrous
7601-89-0
ClNaO4
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
[O-]Cl(=O)(=O)=O.[Na+]
122.436
122.46
2.50g/mL
For analysis ACS
98.0 to 102.0%
MFCD00003480
BAZAXWOYCMUHIX-UHFFFAOYSA-M
sodium;perchlorate
522606
Crystalline
100g

Chemical Identifiers

7601-89-0
122.436
BAZAXWOYCMUHIX-UHFFFAOYSA-M
522606
[O-]Cl(=O)(=O)=O.[Na+]
ClNaO4
MFCD00003480
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
sodium;perchlorate
Safety and Handling

Safety and Handling

EINECSNumber : 231-511-9

Documents