Please login to your online account to display your discounted pricing

Sodium perchlorate, +99%, for analysis, ACROS Organics™


Quantity 5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC197120051

 View more versions of this product

Catalog No. AC197120051

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 122.44
Physical Form Crystals or Crystalline Powder
Assay ≥99.0% (on dry substance)
Quantity 5kg
Color White
Purity Grade For analysis
pH 6.0 to 8.0 (5% aq. soln.)
Heavy Metals (as Pb) 5ppm max.
Loss on Drying 2.0% max. (130°C, 2 hrs)
Packaging Plastic bottle
Linear Formula NaClO4
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause fire or explosion; strong oxidizer.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Sodium perchlorate
Molecular Formula ClNaO4
CAS 7601-89-0
Assay Percent Range 99+%
Calcium (Ca) 200ppm max.
Potassium (K) 500ppm max.
Insoluble in Water 0.005% max.
Iron (Fe) 5ppm max.
Sulfate 20ppm max.
Chlorine (Cl) 30ppm max.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Sodium perchlorate
CAS 7601-89-0
Purity Grade For analysis
Assay ≥99.0% (on dry substance)
Assay Percent Range 99+%
Molecular Formula ClNaO4
Linear Formula NaClO4
Formula Weight 122.44
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause fire or explosion; strong oxidizer.

SDS & Safety Documents