missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium perchlorate, 98%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 342180025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Sodium perchlorate | |
7601-89-0 | |
Liquid | |
98% | |
NaClO4 | |
15, 8784 | |
Solubility in water: 209g/100mL (15°C). Other solubilities: 170g/100mL water (0°C),300g/100mL water (80°C),soluble in alcohol | |
[O-]Cl(=O)(=O)=O.[Na+] | |
122.44 | |
122.44 | |
Extra Pure | |
Plastic bottle |
482.0°C | |
Colorless to Yellow | |
2.5 kg | |
ClNaO4 | |
MFCD00003480 | |
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian | |
BAZAXWOYCMUHIX-UHFFFAOYSA-M | |
sodium;perchlorate | |
522606 | |
98% | |
2 % max. (130°C, 2 hrs) |
Chemical Identifiers
7601-89-0 | |
122.44 | |
BAZAXWOYCMUHIX-UHFFFAOYSA-M | |
522606 | |
[O-]Cl(=O)(=O)=O.[Na+] |
ClNaO4 | |
MFCD00003480 | |
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian | |
sodium;perchlorate |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
GHS Signal Word: Danger
EINECSNumber : 231-511-9
RUO – Research Use Only