missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium percarbonate, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC370730100

 View more versions of this product

Catalog No. AC370730100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥13% (available oxygen)
Granular Powder
White
Plastic bucket
MFCD00043204
1·5 H2O2
157.01
10kg
NaCO3·1.5H2O
Sodium percarbonate
Sodium carbonate hydrogen peroxide
15630-89-4

Chemical Identifiers

1·5 H2O2
157.01
Sodium carbonate hydrogen peroxide
≥13% (available oxygen)
NaCO3·1.5H2O
MFCD00043204
Sodium percarbonate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger