missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium percarbonate, 13-14% active oxygen, Acros Organics

Manufacturer:  Thermo Fisher Scientific AC370735000

 View more versions of this product

Catalog No. AC370735000



Description

Description

Oxidation reagent
Specifications

Specifications

Sodium percarbonate
1·5 H2O2
MFCD00043204
Solubility in water: 150g/L.
157.01
≥13% (available oxygen)
Solubility in water: 150g/L.
500g
15630-89-4
1·5 H2O2
Sodium carbonate hydrogen peroxide
157.01
Granular Powder
Plastic bottle
White

Chemical Identifiers

15630-89-4
157.01
Sodium carbonate hydrogen peroxide
1·5 H2O2
MFCD00043204
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 239-707-6

SDS Documents