missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium percarbonate, 13-14% active oxygen, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A160450E

 View more versions of this product

Catalog No. AAA160450E



Description

Description

Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong reducing agents, strong acids, organic materials and powdered metals.
Specifications

Specifications

Sodium percarbonate
15630-89-4
MFCD00043204
UN1479
Hydrogen peroxide-Sodium carbonate adduct
157.01
13 to 14% active oxygen
Na2CO3·1.5H2O2
2500 g
Moisture sensitive
Soluble in water.

Chemical Identifiers

15630-89-4
Na2CO3·1.5H2O2
MFCD00043204
Hydrogen peroxide-Sodium carbonate adduct
Safety and Handling

Safety and Handling

GHS H Statement
H272-H318
May intensify fire; oxidizer.
Causes serious eye damage.

P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P370+P378q-P501c

H272-H302-H318

DOTInformation : Hazard Class: 5.1; Packaging Group: III

EINECSNumber : 239-707-6

RTECSNumber : FG0750000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only