Please login to your online account to display your discounted pricing

Sodium perborate monohydrate, 95%, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC386412500

 View more versions of this product

Catalog No. AC386412500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94% (active oxygen)
Physical Form Crystalline Powder
Formula Weight 99.81
Quantity 250g
Color White
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
May damage the unborn child. Suspected of damaging fertility.
May cause respiratory irritation.
Causes serious eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.
CAS 10332-33-9
MDL Number MFCD00149230
Chemical Name or Material Sodium perborate monohydrate
Assay Percent Range 95%
Molecular Formula BNaO3·H2O
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Obtain special instructions before use.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Sodium perborate monohydrate
CAS 10332-33-9
Assay ≥94% (active oxygen)
Assay Percent Range 95%
Molecular Formula BNaO3·H2O
Formula Weight 99.81
MDL Number MFCD00149230
Accessories

Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Obtain special instructions before use.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May damage the unborn child. Suspected of damaging fertility.
May cause respiratory irritation.
Causes serious eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.

SDS & Safety Documents