Learn More
Sodium pentacyanonitrosylferrate(III) dihydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals
For Chromatographic Detection of Peptides
Supplier: Thermo Scientific Chemicals 08768330
Description
For chromatographic detection of peptides.Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium pentacyanonitrosylferrate(III) dihydrate | |
Red | |
250 g | |
C5H4FeN6Na2O3 | |
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
JFPDVRGGFBUORC-UHFFFAOYSA-N | |
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate | |
11953895 | |
ACS | |
Hygroscopic |
13755-38-9 | |
Crystals | |
99.0-102.0% | |
MFCD00149192 | |
Soluble in water. Slightly soluble ethanol. | |
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] | |
297.95 | |
297.95 (261.92 Anhydrous) | |
Odorless | |
1.72 |
Chemical Identifiers
13755-38-9 | |
297.95 | |
JFPDVRGGFBUORC-UHFFFAOYSA-N | |
11953895 | |
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] |
C5H4FeN6Na2O3 | |
MFCD00149192 | |
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate |
Safety and Handling
P260-P264b-P270-P271-P301+P310-P304+P340-P312-P314-P330-P501c
H301
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 238-373-9
RTECSNumber : LJ8925000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only