Please login to your online account to display your discounted pricing

Sodium Oxalate, For Nitrogen (Nitrite), Certified, 0.0250M ±0.0005M (0.05N), LabChem™

$62.89 - $62.89

Chemical Identifiers

Chemical Name or Material Sodium Oxalate
CAS 7732-18-5
CAS Max % 99.66
CAS 62-76-0
CAS Max % 0.34
Name Note For Nitrogen (Nitrite)
Purity Grade Certified Grade
Linear Formula Na2C2O4
Formula Weight 134
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC247201

 
lab chem
LC247201
500mL Each for $62.89
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 134
Chemical Name or Material Sodium Oxalate
Traceability to NIST Traceable to NIST
Decomposition Information Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 500mL
Name Note For Nitrogen (Nitrite)
Physical Form Liquid
Concentration 0.0250M ±0.0005M (0.05N)
Linear Formula Na2C2O4
Density 1g/mL
CAS 7732-18-5
CAS Max % 99.66
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 62-76-0
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 0.34
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.