missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium oxalate, ACS reagent, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446965000

 View more versions of this product

Catalog No. AC446965000



Description

Description

Specifications

Specifications

Sodium oxalate
C2Na2O4
0.002% max
disodium oxalate, ethanedioic acid disodium salt, ethanedioic acid, disodium salt, natriumoxalat, natriumoxalat german, oxalic acid disodium salt, oxalic acid, disodium salt, sodium oxalate, stavelan sodny czech, unii-7u0v68lt9x
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
disodium;oxalate
6125
ACS Reagent
0.005% max
Solubility in water: 37g/L (20°C). Other solubilities: 6.3 g/100mL water (100°C), insoluble in ethanol and ether
250.0°C to 270.0°C
62-76-0
NaO2CCO2Na
15,8781
Solubility in water: 37g/L (20°C). Other solubilities: 6.3g/100mL water (100°C),insoluble in ethanol and ether
C(=O)(C(=O)[O-])[O-].[Na+].[Na+]
134
134
0.002% max
Plastic bottle
0.002% max
500g

Chemical Identifiers

62-76-0
134
disodium oxalate, ethanedioic acid disodium salt, ethanedioic acid, disodium salt, natriumoxalat, natriumoxalat german, oxalic acid disodium salt, oxalic acid, disodium salt, sodium oxalate, stavelan sodny czech, unii-7u0v68lt9x
disodium;oxalate
C2Na2O4
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
6125
C(=O)(C(=O)[O-])[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 200-550-3

SDS Documents