Please login to your online account to display your discounted pricing

Sodium Oxalate, ACS Grade, ≥ 99.5%, LabChem™

$212.89 - $212.89

Chemical Identifiers

Chemical Name or Material Sodium Oxalate
CAS 62-76-0
CAS Max % 100
Purity Grade ACS Grade
Linear Formula Na2C2O4
Formula Weight 134
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC247101

 
lab chem
LC247101
500g Each for $212.89
Description & Specifications

Specifications

Assay ≥99.5%
Formula Weight 134
Chemical Name or Material Sodium Oxalate
EINECS Number 200-550-3
Identification Passes Test
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Powder
Solubility Soluble in water
Specific Gravity 2.34g/cm3
Storage Information Room Temperature
Linear Formula Na2C2O4
CAS 62-76-0
Decomposition Information Carbon monoxide; Carbon dioxide
Color White
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed or in contact with skin.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.
Density 2.34g/cm3
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed or in contact with skin.
Causes serious eye irritation.