missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium oxalate, 99%

Manufacturer:  Alfa Aesar™ A1164822

 View more versions of this product

Catalog No. AAA1164822


 

Description

Description

Applications
Sodium oxalate is used as reducing agent, as a primary standard for standardizing potassium permanganate solutions and as a neutralizer for exothermic acidic reactions. It finds application in metal cleaning preparations, metal extraction and separation, blueprint coatings, electroplating baths, special cement manufacturing, plant nutrition as well as in textile and pharmaceutical industries. It plays an important role to remove calcium ions from blood plasma and prevents blood clotting.

Solubility
Soluble in water and formic acid. Insoluble in ether and alcohol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Sodium oxalate
99%
MFCD00012465
14,8650
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
disodium;oxalate
6125
~250°C to 270°C (decomposition)
Odorless
62-76-0
C2Na2O4
3631622
Oxalic acid sodium salt
C(=O)(C(=O)[O-])[O-].[Na+].[Na+]
133.998
134
100g

Chemical Identifiers

62-76-0
133.998
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
6125
C2Na2O4
MFCD00012465
Oxalic acid sodium salt
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : KI1750000

TSCA : TSCA

SDS Documents