Learn More
Sodium oxalate, 99.5%, for analysis, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC270540051
Specifications
20ppm max | |
20ppm max | |
100ppm max | |
250.0°C to 270.0°C | |
7 to 8.5 (2.5% soln. at 20°C) | |
62-76-0 | |
C2Na2O4 | |
MFCD00012465 | |
disodium oxalate, ethanedioic acid disodium salt, ethanedioic acid, disodium salt, natriumoxalat, natriumoxalat german, oxalic acid disodium salt, oxalic acid, disodium salt, sodium oxalate, stavelan sodny czech, unii-7u0v68lt9x | |
ZNCPFRVNHGOPAG-UHFFFAOYSA-L | |
disodium;oxalate | |
6125 | |
Fine Powder | |
Analysis |
Sodium oxalate | |
0.05% max. (105°C) | |
White | |
5kg | |
p.a. | |
99.5% | |
NaO2CCO2Na | |
15,8781 | |
Solubility in water: 37g/L (20°C). Other solubilities: 6.3 g/100mL water (100°C), insoluble in ethanol and ether | |
C(=O)(C(=O)[O-])[O-].[Na+].[Na+] | |
134 | |
134 | |
99.5% |
Chemical Identifiers
62-76-0 | |
134 | |
ZNCPFRVNHGOPAG-UHFFFAOYSA-L | |
6125 | |
C(=O)(C(=O)[O-])[O-].[Na+].[Na+] |
C2Na2O4 | |
MFCD00012465 | |
disodium oxalate, ethanedioic acid disodium salt, ethanedioic acid, disodium salt, natriumoxalat, natriumoxalat german, oxalic acid disodium salt, oxalic acid, disodium salt, sodium oxalate, stavelan sodny czech, unii-7u0v68lt9x | |
disodium;oxalate |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-550-3
RTECSNumber : KI1750000
TSCA : TSCA