Please login to your online account to display your discounted pricing

Sodium oxalate, 98.5%, extra pure, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC207720025

 View more versions of this product

Catalog No. AC207720025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0% (KMnO4 titration)
Formula Weight 134
Purity Grade Extra Pure
Physical Form Fine Powder
Quantity 2.5kg
Color White
TSCA TSCA
EINECS Number 200-550-3
RTECS Number KI1750000
Merck Index 15, 8781
Iron (Fe) 50ppm max.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Health Hazard 1 Warning
Loss on Drying 0.1% max. (105°C, 2 hrs)
Packaging Plastic bottle
Additional Information passing through 125μm sieve:> 95%
Synonym Oxalic acid, sodium salt
Insoluble in Water 0.2% max.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula NaO2CCO2Na
CAS 62-76-0
Chemical Name or Material Sodium oxalate
Assay Percent Range 98.5%
Molecular Formula C2Na2O4
Name Note pure
MDL Number MFCD00012465

Specifications

Chemical Name or Material Sodium oxalate
Name Note pure
CAS 62-76-0
Purity Grade Extra Pure
Assay ≥98.0% (KMnO4 titration)
Assay Percent Range 98.5%
Molecular Formula C2Na2O4
Linear Formula NaO2CCO2Na
Formula Weight 134
MDL Number MFCD00012465
Synonym Oxalic acid, sodium salt
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.

SDS & Safety Documents