Please login to your online account to display your discounted pricing

Sodium Oxalate


Quantity 25g
Packaging Reagent Grade

Manufacturer: fisher science education  SS0720-25G

 View more versions of this product

Catalog No. S93375

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 134
Quantity 25g
Color Colorless
DOT Information DOT Class 6.1, : Poison
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Chemical Name or Material Sodium Oxalate
Shelf Life 12 months
Packaging Reagent Grade
Linear Formula Na2C2O4
ChemAlert Storage Symbol White
CAS 62-76-0

Specifications

Chemical Name or Material Sodium Oxalate
CAS 62-76-0
Linear Formula Na2C2O4
Formula Weight 134
Accessories
 

12 months