Please login to your online account to display your discounted pricing

Sodium Oxalate

$7.10 - $7.10

Chemical Identifiers

Chemical Name or Material Sodium Oxalate
CAS 62-76-0
Linear Formula Na2C2O4
Formula Weight 134
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S93375

 
fisher science education
SS0720-25G
Reagent Grade 25g Each for $7.10
Description & Specifications

Specifications

Formula Weight 134
Quantity 25g
Color Colorless
DOT Information DOT Class 6.1, : Poison
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Chemical Name or Material Sodium Oxalate
Shelf Life 12 months
Packaging Reagent Grade
Linear Formula Na2C2O4
ChemAlert Storage Symbol White
CAS 62-76-0
Safety and Handling
 

12 months